Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S100Z Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584092
Description
Immunogen sequence: MPTQLEMAMD TMIRIFHRYS GKARKRFKLS.
Members of the S100 protein family contain 2 calcium-binding EF-hands and exhibit cell-type specific expression patterns. For additional background information on S100 proteins, see MIM 114085.
Specifications
S100Z | |
Polyclonal | |
Unconjugated | |
S100Z | |
Gm625; Protein S100-Z; S100 calcium binding protein Z; S100 calcium binding protein, zeta; S100 calcium-binding protein Z; S100Z; S100-zeta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170591 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WXG8 | |
S100Z | |
Recombinant protein corresponding to Human S100Z. Recombinant protein control fragment (Product #RP-108392). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction