Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ MMB-FUBINACA Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco S1111ML
Description
MMB-FUBINACA, also known as AMB-FUBINACA and FUB-AMB, is a potent indazole-based synthetic cannabinoid receptor agonist sold as a designer drug. This certified Snap-N-Spike™ solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications. For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.Specifications
75-05-8, 1971007-92-7 | |
UN 1648 3 / PGII | |
383.42 | |
1 mL |
C21H22FN3O3 | |
100 μg/mL in acetonitrile | |
Ampule | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?
Provide Content Correction