Learn More
Thermo Scientific™ S120-VT Vertical Rotor Available on GSA/VA Contract for Federal Government customers only.
Support a wide variety of applications with this eight-place capacity rotor capable of 120,000rpm and 500,237 x g.
Manufacturer:  Thermo Scientific™ 45593
Catalog No. 45-593
Add to cart
Description
Description
Support a wide variety of applications with Thermo Scientific™ S120-VT Vertical Rotor, an eight-place capacity rotor capable of 120,000rpm and 500,237 x g.
Includes:
  • 2mL Polyallomer Re-SealTM Tube (2 pkgs. of 50)
  • Crown (8 pkgs. of 1)
  • Plugs (1 pkg. of 10)
  • Wrench, rotor cap wrench, and torque wrench
  • Removal tool
  • Rotor cap (8 pkgs. of 1)
  • Instruction manual
  • Rotor holding fixture
  • Vacuum grease
  • Lubricant
Compatible with:
Thermo Scientific™ Sorvall™ MX Series Floor Model Micro-Ultracentrifuge, MTX 150 Series Micro-Ultracentrifuge
Recommended for:
Small volume isopycnic banding or CsCI/DNA isolation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
S120-VT Vertical Rotor
Fixed Angle Rotor
500,237 x g
31.1mm
7.9
11mm
Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges
410,162 x g
Titanium
8 x 2mL
120,000rpm
19.9mm
32mm
1.4 lb. (0.63kg)
320,088 x g