Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P3/EDG-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224762SS
Description
S1P3/EDG-3 Polyclonal specifically detects S1P3/EDG-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S1P3/EDG-3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human and 95% homologous in mouse. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Q99500 | |
S1PR3 | |
A portion of amino acids 300-350 of human Sphingolipid Receptor Edg3/S1P3 was used as the immunogen. | |
0.025 mg | |
Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
1903 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction