Learn More
S1P3/EDG-3 Mouse anti-Human, Alexa Fluor 405, Clone: 776808, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7524V100UG
Catalog No. FAB7524V100
Add to cart
This item is not returnable. View return policy
Description
Description
S1P3/EDG-3 Monoclonal antibody specifically detects S1P3/EDG-3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
S1P3/EDG-3
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q99500
S1PR3
Human S1P3/EDG-3 transfected NS0 cells Accession # Q99500
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human S1P 3EDG-3 in direct ELISAs.
Flow Cytometry
776808
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
EDG3, EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, S1PR3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3
Mouse
IgG2a
RUO
Primary
1903
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)