Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P4/EDG-6, Mouse anti-Human, Clone: 1012512, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $298.50

Specifications

Antigen S1P4/EDG-6
Clone 1012512
Immunogen NS0 mouse myeloma cell line transfected with human S1P4/EDG-6, Met1-Ile384, Accession # O95977
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10321100
View Documents
R&D Systems
MAB10321100
100 ug
Each for $298.50
Add to cart
 
MAB10321SP
View Documents
R&D Systems
MAB10321SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

S1P4/EDG-6 Monoclonal antibody specifically detects S1P4/EDG-6 in Human Samples. It is validated for Flow Cytometry, CyTOF.
Specifications

Specifications

S1P4/EDG-6
NS0 mouse myeloma cell line transfected with human S1P4/EDG-6, Met1-Ile384, Accession # O95977
Primary
Human
Unconjugated
8698
Purified
Flow Cytometry, CyTOF
RUO
1012512
Mouse
Monoclonal
IgG2b
EDG6, EDG6S1P receptor Edg-6, Endothelial differentiation G-protein coupled receptor 6, endothelial differentiation, G-protein-coupled receptor 6, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor6,endothelial differentiation, G protein coupled receptor 6, S1P receptor 4, S1P4LPC1, S1PR4, SLP4, sphingosine 1-phosphate receptor 4, Sphingosine 1-phosphate receptor Edg-6, sphingosine-1-phosphate receptor 4
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit