Learn More
S1P5/EDG-8 Rabbit anti-Mouse, Alexa Fluor 700, Clone: 1196A, R&D Systems™
Rabbit MonoclonalAntibody
Manufacturer:  R&D Systems FAB9084N100UG
Catalog No. FAB9084N100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
S1P5/EDG-8
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Edg-8, EDG8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, LPB4, S1P receptor 5, S1P5, S1PR5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Mouse S1P 5EDG-8 peptide corresponding to the N-terminal extracellular sequence Accession # Q91X56
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse S1P 5EDG-8 in Western blots.
Flow Cytometry
1196A
Flow Cytometry 0.25-1 ug/10^6 cells
Q91X56
Rabbit
IgG
RUO
Primary
53637
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)