Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ S26948

Selective PPARgamma agonist; antidiabetic agent

Manufacturer:  Tocris™ 3690/50

 View more versions of this product

Catalog No. 36-905-0R


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Selective PPARγ agonist (EC50 = 8.83nM). Drives normal adipocyte differentiation and exhibits a lipid-lowering effect; reduces atherosclerosis in E2-KI mice. Displays antidiabetic properties and improves hepatic insulin sensitivity in intralipid-infusion (IL) rats.
  • Chemical Name: [[4-[2-(6-Benzoyl-2-oxo-3(2H)-benzothiazolyl)ethoxy]phenyl]methyl]-1,3-propanedioic acid dimethyl ester
  • Formula: C28H25NO7S
  • CAS Number: 353280-43-0
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

S26948
>99
50mg
S26948
519.57
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit