Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ S3QEL 2

Suppressor of complex III superoxide production

$67.50 - $285.50

Specifications

Chemical Name or Material 1-(3,4-Dimethylphenyl)-N,N-dipropyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Formula C19H25N5
CAS 890888-12-7
Product Type Target: Additional ROS Donor Products
Formula Weight 323.44
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-351-0 View Documents Tocris Bioscience™
5735/10
10mg Each for $67.50
Add to cart
 
57-355-0 View Documents Tocris Bioscience™
5735/50
50mg Each for $285.50
Add to cart
 
Description

Description

Suppresses superoxide production by complex III in the mitochondria without affecting normal electron flux or cellular oxidative phosphorylation (IC50 = 1.7 μM). Suppresses HIF-1α accumulation and enhances survival and function of primary pancreatic islets in vitro. Cell permeable.
Specifications

Specifications

1-(3,4-Dimethylphenyl)-N,N-dipropyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
890888-12-7
323.44
C19H25N5
Target: Additional ROS Donor Products
Safety and Handling

Safety and Handling

Recommended Storage : Store at −20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit