Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH3TC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550135
Description
The antibody detects endogenous levels of total S3TC1 / SH3TC1 protein.
SH3TC1 Polyclonal antibody specifically detects SH3TC1 in Human samples. It is validated for Western Blot.Specifications
SH3TC1 | |
Polyclonal | |
Unconjugated | |
SH3TC1 | |
SH3 domain and tetratricopeptide repeat-containing protein 1; SH3 domain and tetratricopeptide repeats 1; SH3 domain and tetratricopeptide repeats-containing protein 1; SH3TC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54436 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8TE82 | |
SH3TC1 | |
Synthesized peptide derived from internal of human SH3TC1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction