Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S5a/Angiocidin Mouse anti-Human, Clone: CPTC-PSMD4-3, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen S5a/Angiocidin
Clone CPTC-PSMD4-3
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Protein Array
Applications Western Blot, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121974
View Documents
Novus Biologicals
NBP30725020UG
20 μg
Each for $138.37
Add to cart
 
NB121973
View Documents
Novus Biologicals
NBP30725010UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

S5a/Angiocidin Monoclonal antibody specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Protein Array
Specifications

Specifications

S5a/Angiocidin
0.2 mg/ml
Western Blot, Peptide Array
Purified
RUO
Human
5710
IgG2b Kappa
Primary
Store at 4°C.
CPTC-PSMD4-3
Western Blot 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Neuroscience, Ubiquitin Proteasome Pathway
10 mM PBS with 0.05% BSA
Recombinant full-length human S5a/Angiocidin protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit