Learn More
Thermo Scientific™ SA-600 Fixed Angle Rotor Available on GSA/VA Contract for Federal Government customers only.
Process samples efficiently with the 12-place, autoclavable SA-600 12 x 50mL Fixed-Angle Rotor.
Manufacturer:  Thermo Scientific™ 28500
Catalog No. 28-500
Add to cart
Description
Description
Process samples efficiently with the 12-place, autoclavable Thermo Scientific™ SA-600 Fixed-Angle Rotor.
Compatible with:
Thermo Scientific™ Sorvall™ Evolution RC, RC-6 Plus centrifuge series
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SA-600 Fixed Angle Rotor
Fixed Angle Rotor
41,837 x g
129.6mm
747
29mm
Thermo Scientific™ Sorvall™ RC 6 Plus and Evolution RC superspeed Centrifuges
Biocontainment Certified by CAMR, HPA, Porton Down, U.K.
Aluminum
12 x 50mL
17,000rpm
55.2mm
108mm
22.3 lb. (10.1kg)
34°
17,819 x g