Learn More
SA1 Mouse anti-Human, DyLight 405, Clone: 8810, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243750V
Catalog No. NBP243750V
Add to cart
Description
Description
SA1 Monoclonal antibody specifically detects SA1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SA1
8810
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
STAG1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA1. The exact sequence is proprietary.
Protein A purified
Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary
10274
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human