Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SA1 Mouse anti-Human, FITC, Clone: 8810, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243750F

Catalog No. NBP243750F


Add to cart

Description

Description

SA1 Monoclonal antibody specifically detects SA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SA1
8810
Immunocytochemistry/Immunofluorescence
Purified
STAG1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SA1. The exact sequence is proprietary.
Protein A purified
Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary
10274
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit