Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAA Goat anti-Horse, Polyclonal , Abnova™

Goat polyclonal antibody raised against horse SAA.

Manufacturer:  Abnova Corporation PAB29721

Catalog No. 89-934-895

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

SAA
Unconjugated
ELISA (2 ug/mL) Immunoprecipitation (2 ug/mg) Western Blot (0.05-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to horse SAA.
Antigen Affinity Purification
RUO
Primary
Equine
ELISA, Immunoprecipitation, Western Blot
Goat polyclonal antibody raised against horse SAA.
In PBS, pH 7.2 (0.09% sodium azide)
Goat
IgG
100 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit