Learn More
SAA Mouse anti-Human, Mouse, Clone: Reu86.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA533370
Catalog No. PIMA533370
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody reacts with the SAA-1 type.
The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
Specifications
Specifications
SAA
Monoclonal
0.5 μg/mL
PBS with no preservative
P05366, P0DJI8
MGC111216, PIG4, SAA, TP53I4
Mouse
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Reu86.5
Unconjugated
SAA1
liquid
SAA1
IgG1
0.5 mg
4° C
Primary
20208, 6288