Learn More
SAA3 Rat anti-Mouse, Alexa Fluor™ 647, Clone: JOR110A, BD Pharmingen™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566652
Catalog No. BDB566652
Add to cart
Description
Description
SAA3 Monoclonal antibody specifically detects SAA3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
SAA3
JOR110A
Alexa Fluor 647
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2b, κ
RUO
Primary
SAA3