Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAAL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | SAAL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157059
|
Novus Biologicals
NBP157059 |
100 μL |
Each of 1 for $436.00
|
|
Description
SAAL1 Polyclonal specifically detects SAAL1 in Human samples. It is validated for Western Blot.Specifications
SAAL1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ41463, protein SAAL1, serum amyloid A-like 1 | |
SAAL1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q96ER3 | |
113174 | |
Synthetic peptides corresponding to SAAL1 (serum amyloid A-like 1) The peptide sequence was selected from the middle region of SAAL1. Peptide sequence EKLMLEWVRNGAAQPLDQPQEESEEQPVFRLVPCILEAAKQVRSENPEWL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title