Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
81-07-2
C7H5NO3S
183.18
CVHZOJJKTDOEJC-UHFFFAOYSA-N
saccharin,saccharine,o-sulfobenzimide,o-benzoic sulfimide,saccharimide,benzosulfimide,o-benzosulfimide,benzoic sulfimide,benzosulphimide,saccharinose
5143
CHEBI:32111
1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)NS2(=O)=O
81-07-2
C7H5NO3S
183.18
CVHZOJJKTDOEJC-UHFFFAOYSA-N
saccharin,saccharine,o-sulfobenzimide,o-benzoic sulfimide,saccharimide,benzosulfimide,o-benzosulfimide,benzoic sulfimide,benzosulphimide,saccharinose
5143
CHEBI:32111
1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)NS2(=O)=O
Sulfated Ash 0.2% max.
Infrared Spectrum Authentic
Melting Point 226.0°C to 230.0°C
Loss on Drying 1% (105°C, 2 hrs) max.
Packaging Plastic bottle
Color White
Quantity 2.5kg
Assay Percent Range 98+%
Beilstein 27,168
Merck Index 15,8445
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g/31mL alcohol - 1g/12mL acetone, 1g/50mL glycerol, soluble in amyl acetate and ethyl acetate, slightly soluble in chloroform and ether, soluble in dil. solution of ammonia
Formula Weight 183.18
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material Saccharin
Show More Show Less