Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Calcium, Crystal, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
100 lbs.
12 kg
125 g
2.5 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6381-91-5
C14H10CaN2O7S2
422.44
XOCXGVIOTKFUSU-UHFFFAOYSA-L
calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12
6381-91-5
C14H10CaN2O7S2
422.44
XOCXGVIOTKFUSU-UHFFFAOYSA-L
calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12

CA167, 6381-91-5, C14H8CaN2O6S2•31/2H2O

Spectrum™ Saccharin Calcium, Crystal, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 99 to 101%
Packaging Poly Pail
Water 0.15
Quantity 12 kg