Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Sodium, Dihydrate, Crystalline, USP, 99-101%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6155-57-3
C7H8NNaO5S
241.19
NAUHOSQQKJTUAX-UHFFFAOYSA-M
sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
6155-57-3
C7H8NNaO5S
241.19
NAUHOSQQKJTUAX-UHFFFAOYSA-M
sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate dihydrate
O.O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

SO195, 6155-57-3, C7H4NNaO3S•2H2O

Spectrum™ Saccharin Sodium, Dihydrate, Crystalline, USP is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Water 0.15
Quantity 500 g
CAS Max % 1
Percent Purity 99 to 101%
Grade USP