Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAE1 Mouse anti-Human, Clone: OTI6B11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526712

Catalog No. PIMA526712


Add to cart

Description

Description

Small ubiquitin-like modifiers (SUMOs) are a family of small, related proteins (SUMO-1/2/3/4) that can be enzymatically attached to a target protein by a post-translational modification process termed sumoylation, a major regulator of protein function in cellular processes such as nuclear transport, transcriptional regulation, apoptosis and protein stability. This sumoylation is effected by the heterodimeric enzyme SAE1/SAE2 and the SUMO-1-conjugating enzyme Ubch9. The sumoylation pathway mediated by SAE1/SAE2 is distinct from other ubiquitin-like protein (Ubl) pathways.
Specifications

Specifications

SAE1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UBE0
SAE1; AOS1; SUA1; HSPC140; SUMO-1 activating enzyme subunit 1; sentrin/SUMO-activating protein AOS1; SUMO-1 activating enzyme E1 N subunit; ubiquitin-like protein SUMO-1 activating enzyme; SUMO1 activating enzyme subunit 1; FLJ3091; UBLE1A; activator of SUMO1; ubiquitin-like 1-activating enzyme E1A
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10055
Immunohistochemistry (Paraffin), Western Blot
OTI6B11
Unconjugated
SAE1
Liquid
SAE1
Full length human recombit protein of SAE1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit