Learn More
Thermo Scientific™ SAF Fixative / Empty Vial Two-Vial Set
Collect, preserve, concentrate and permanently stain intestinal parasites with SAF Fixative plus an empty vial in a convenient two-vial set with color-coded, sporked caps.
Supplier: Thermo Scientific™ R21732
Description
Collect, preserve, concentrate and permanently stain intestinal parasites with a convenient two-vial set. Thermo Scientfic™ SAF Fixative / Empty Vial Two-Vial Set is part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
- Cary Blair with Indicator
- Formalin 10%
- Formalin 5%
- PVA Modified Fixative
- SAF Fixative
- Zinc PVA Fixative
- Empty Vial
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
SAF Fixative / Empty Vial Two-Vial Set | |
15 mL | |
2 Vials/Set. Packaged in zip-seal bags. | |
6 Sets/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.