Learn More
Medisca Safe-Sense™ Blue Nitrile Powder Free Chemo Rated Exam Gloves
Medisca Safe-Sense™ Powder Free Chemo Rated Exam Gloves provide maximum protection for non-sterile hazardous compounding, administration, disposal and other related chemotherapy activities. They are resistant to puncture, abrasion and snag.
$147.50
Specifications
AQL 1.5
Certifications/Compliance ASTM D6978-05
Certifications/Compliance (Europe) EN455
Chemotherapeutic Rated Yes
FDA 510 (k) Yes
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-048-000
View Documents
Medisca
574202
Small
Case of 1000 for $147.50
Add to cart
18-048-001
View Documents
Medisca
574302
Medium
Case of 1000 for $147.50
Add to cart
18-048-002
View Documents
Medisca
574402
Large
Case of 1000 for $147.50
Add to cart
18-048-003
View Documents
Medisca
574502
X-Large
Case of 1000 for $147.50
Add to cart
Description
Description
  • Medical examination gloves
  • Improves user and product protection for superior durability
  • Polyurethane coating for easy donning and removal
  • Beaded cuff to reduce glove roll down and ripping
  • Textured fingertips to provide grip and sensitivity
  • Produced with distilled water for a silky feeling
  • Disposable
  • Meets EN455 standard and ASTM D6319
  • Meets ASTM D6978 resistant to permeation by chemotherapy drugs
  • Manufactured in an ISO 13485 certified facility
  • Meets FDA regulations for food handling applications
Specifications
Specifications
1.5
EN455
Yes
4 mil
ASTM D6978-05
Yes
Healthcare, Laboratory, Industrial
0.10 mm