Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ SafeBasics™ Isolation Gown
Non-surgical Isolation Gown with belt, intended to protect health care patients and personnel from the transfer of microorganisms, body fluids (fluid repellent) and particulate material.
$77.00
Specifications
Material | Spunbond Polypropylene |
---|---|
Disposable | Yes |
Sterility | Non-sterile |
For Use With (Application) | Protection from cross contamination |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Size | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Color | Price | Quantity & Availability | ||||
19-910-553
|
Medicom™
8023 |
Regular | Blue |
Case of 5 for $77.00
|
|
19-910-552
|
Medicom™
8010 |
X-Large | Yellow | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-910-551
|
Medicom™
8009 |
Regular | Yellow | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Comfortable fit with elastic cuffs, back closure, waist and neck ties
- Fluid repellent material
- Superior strength provided with sewn seams
- Made of non-woven fabric
- Belt on the right side
- Nonsterile
- Single use, disposable and latex-free
- 10 per bag, 5 bags per case
Specifications
Spunbond Polypropylene | |
Non-sterile | |
Elastic | |
Long sleeve |
Yes | |
Protection from cross contamination | |
Yes | |
Not Autoclavable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title