Brady™ Safelex™ Lockout Device

Lockout gates, handles, valves and more! The Brady™ Safelex™ Lockout Device is a versatile solution for all of your facility's lockout needs.

Manufacturer: Brady™ 145550

 View more versions of this product

Catalog No. 19-103-929

  Check Availability
Add to Cart

    Specifications

    Description Maximum of 6 padlocks
    Dimensions (L x W) 5 in. x 2.5 in.
    Includes 6 ft. red cable
    Material Polycarbonate body and nylon cable
    Quantity 1/Each
    Color Black/red
    Product Type Non-conductive cable
    Type Hasp integrated
    Width (English) 2.5 in.

    • Provides solutions for locking out gate valves, handles and other oversized devices
    • Versatile solution that is perfect for all of your difficult lock points

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit