Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Safelex™ Lockout Device
Lockout gates, handles, valves and more! The Brady™ Safelex™ Lockout Device is a versatile solution for all of your facility's lockout needs.
Supplier: Brady™ 145553
View more versions of this product
Safety Services Available
Overview
LOTO Assessments
Description
- Provides solutions for locking out gate valves, handles and other oversized devices
- Versatile solution that is perfect for all of your difficult lock points
Specifications
Lockout Cable with Hasp | |
Polycarbonate with Sheathed Steel Cable | |
5 x 2.5 in. | |
1 Ea. |
Black/Red | |
Maximum of 6 padlocks | |
6 ft. steel cable | |
2.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction