Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ SaferGrip™ Waterproof Cryogenic Gloves - Elbow Length

Waterproof Cryogenic Gloves offer thermal protection against liquid nitrogen splashes and other cold temperature tasks. SaferGrip™ silicone-coated palm enhances grip and durability.

$250.80

Specifications

Color Blue
Coating Material Silicone
Description Elbow Length
For Use With (Application) Cryogenic Liquids/Gases
Length (English) 18 in.
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-355-075
View Documents Promotion Details
National Safety Apparel™
G99CRSGPMDEL
Medium
Pair for $250.80
Add to cart
 
17-355-076
View Documents Promotion Details
National Safety Apparel™
G99CRSGPLGEL
Large
Pair for $250.80
Add to cart
 
17-355-077
View Documents Promotion Details
National Safety Apparel™
G99CRSGPXLEL
X-Large
Pair for $250.80
Add to cart
 
Description

Description

  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • SaferGrip™ silicone coated material on the palm for enhanced grip and durability
  • Waterproof, breathable membrane for additional splash protection
  • Made in the USA
Specifications

Specifications

Blue
Elbow Length
18 in.
Nylon
G99CRSGP_ _EL
Silicone
Cryogenic Liquids/Gases
3M™ Thinsulate™
Waterproof Cryogenic Gloves
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit