Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ SaferGrip™ Waterproof Cryogenic Gloves - Shoulder Length
SDP

Catalog Number 17355079
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Waterproof Cryogenic Gloves offer thermal protection against liquid nitrogen splashes and other cold temperature tasks. SaferGrip™ silicone-coated palm enhances grip and durability.

  • Gloves offer protection from ultra-cold environments in a wide variety of applications
  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • SaferGrip™ silicone coated material on the palm for enhanced grip and durability
  • Waterproof, breathable membrane for additional splash protection
  • Made in the USA
Certifications/Compliance (Europe) EN 511
Coated Area Palm
Description Shoulder Length
Coating Material Silicone
Color Blue
Material Nylon
Product Type Waterproof Cryogenic Gloves
Size Large
For Use With (Application) Cryogenic Liquids/Gases
Liner Material 3M™ Thinsulate™
Series G99CRSGP_ _SH
Length (English) 26 in.
Length (Metric) 66 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit