Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Safety Can For Liquid Disposal

Catalog No. p-7070903
Click to view available options
Capacity (English):
2 gal.
5 gal.
Color:
Red
White
Height (English):
14.8 in.
20 in.

Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.

High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.

Specifications

Product Type Safety Can
Description Safety Can for Liquid Disposal
Capacity (English) 5 gal.
Color White
For Use With (Application) Liquid Disposal
Material HDPE
Includes Stainless steel handle, faucet and flame arrester
Height (English) 20 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.