Learn More
Safety Cans Products for Science Education
Safe means of carrying and storing flammable liquids.
$53.50
Specifications
Certifications/Compliance UL, FM, ULC, OSHA, NFPA 30
Finish Powder coat
For Use With (Application) To store Acetone, Toluene, Xylene, Naphtha, Gasoline and other noncorrosive flammable liquids
Includes Screen
Product Type Metallic Can
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
S41789 Products for Science Education
View Documents
Eagle Manufacturing
UI10S
1 gal. 3.8L
Each for $53.50
Add to cart
Description
Description
  • Constructed of galvanized steel with a brass pour spout
  • Single-walled, brass flame arrestor located within the pour spout
  • Store acetone, toluene, xylene, naphtha, gasoline, and other noncorrosive flammable liquids
Compliance
UL listed; FM approved
Specifications
Specifications
UL, FM, ULC, OSHA, NFPA 30
To store Acetone, Toluene, Xylene, Naphtha, Gasoline and other noncorrosive flammable liquids
Metallic Can
Galvanized Steel
Powder coat
Screen
Red