Learn More
Safety Choice™ Single-Dip PVC Gloves
Interlock lined
 View more versions of this product
Catalog No. 19-066-864
Add to cart
Description
Description
Ideal for petrochemical and refining; and handling oils, acids, caustics and alcohols.
  • Available with smooth or semi-rough finish
  • Semi-rough finish provides enhanced wet grip and improved cut-resistance
  • Color: Black
Compliance
Meets EN 388 standards; CE marked.
Specifications
Specifications
Semi rough
mining, construction, refining, Petrochemicals and handling Oily materials
PVC
Black
30.5 cm
Gauntlet
Interlock knit
Men's
12 in.