Learn More
AccuTec Blades Safety Dispenser Cases
AccuTec Blades Safety Dispenser Cases protect users by reducing their exposure to single, and double edge blades
Supplier: AccuTec Blades 940464
Description
- Easy access for safety case
- Positive cap closure to reduce spills
- Degreased blade for reduced oil and particulates
- 3-Facet blade edge for increased sharpness
- High quality stainless steel
Specifications
Carbon Steel | |
1.50 in. | |
Single Edge Blade | |
Aluminum |
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications | |
38 mm | |
Non-sterile | |
600/Cs. |