Learn More
AccuTec Blades Safety Dispenser Cases
AccuTec Blades Safety Dispenser Cases protect users by reducing their exposure to single, and double edge blades
$920.50 - $1112.00
Specifications
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Length (English) Blade 1.50 in.
Length (Metric) Blade 38 mm
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
17-989-152
View Documents
AccuTec Blades
940464
Single Edge Blades
Case of 600 for $1,112.00
Add to cart
17-989-156
View Documents
AccuTec Blades
9509130000
Double Edge Blades
Case for $920.50
Add to cart
Description
Description
  • Easy access for safety case
  • Positive cap closure to reduce spills
  • Degreased blade for reduced oil and particulates
  • 3-Facet blade edge for increased sharpness
  • High quality stainless steel
Specifications
Specifications
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
1.50 in.
38 mm
Non-sterile