Learn More
Restek™ Safety Exhaust Filter with Indicator
SDP
Breakthrough indicator changes from light pink to dark pink when filter is ready to be changed
Supplier:  Restek™ 27833
Catalog No. 06-611-814
Only null left
Add to Cart
Description
Description
  • Safety filter protects lab from volatile compounds in waste bottles
  • 1/4-28 threads for use on Mobi-Cap or Hub-Cap Bottle Tops
  • Replace filter every 3 months or when indicator changes color
  • Filter should be changed more frequently with rigorous use or use in harsh environments
Specifications
Specifications
Mobile Phase Bottle Tops
Indicator
1/4-28
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.