Learn More
Chemical Splash Safety Goggles
Protective eyewear to prevent particulates, water or chemicals from striking the eyes.
This item is not returnable.
View return policy
Description
- Anti-fog lens; double-sided reinforced
- Provide chemical splash protection
- Prevent common virus, pollutant or other particulates from striking the eyes
- Indirect vent and transparent body design
- Material: PVC
- Headband: elastic
Technical Performance:
- Meet ANSI Z87.1 D3 standard for chemical splash protection
- Meets ANSI/ISEA Z87.1 (Optical Quality, Droplet and Splash Hazard, Drop Ball Impact Resistance, Ignition Pass, Minimum Coverage Area)
- Meets GB14866 - Protection against chemical droplets
- Tested for ANSI/ISEA Z87.1-2015, GB 14866-2006. CE and FDA Certified
Specifications
Anti-fog | |
Safety goggles | |
Indirect | |
Double-sided reinforced |
Meet ANSI Z87.1 D3 Standard for Chemical Splash. Tested for ANSI/ISEA Z87.1-2015, CE and FDA Certified. | |
Elastic | |
PVC |