Learn More
3M™ Safety and Inspection Manager Software, Managed Asset Block
3M™ Safety and Inspection Manager Software, Managed Asset Block offers access to real-time data to enhance worker protection. Bocks of additional Managed Assets allow users to right-size service level based on business and operational needs.
Manufacturer:  3M™ 6010002209
Catalog No. 19-071-135
Add to cart
Description
Description
  • Optional add-on for any service package to increase service level for management of an additional 250 assets and 7,500 inspections
  • Enhance Compliance Processes - gain easier access to audit information, which means greater assurance of worker and workplace compliance
  • Save Time - digital inspection management reduces paperwork and time spent on documentation and compliance tasks
  • Be Proactive - on-demand reporting provides a direct line of sight on selected compliance metrics in real-time, helping users gain potentially valuable safety insights
  • Cloud-based for simple deployment and easier maintenance
  • 3M Catalog# IAM-A250
Specifications
Specifications
Connected Safety
Software
Safety