Learn More
Safety Solutions Flu PPE Kits
Helps protect first responders, first receivers, and the private sector from exposure to influenza1
Specifications
Certifications/Compliance | CDC/NIOSH |
---|---|
For Use With (Application) | Helps protect first responders, first receivers, and the private sector from exposure to influenza1 |
Includes:
- One Pair Safety Goggles
- 3M N95 Mask
- Two disposable pairs of Nitrile Gloves
- NexGen Coverall
- One pair Latex Boot Covers
- Two DuPont Broad Spectrum Wipes
- Biohazard Bag
- Donning/Doffing Instruction Card
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
19-160-970
![]() |
Safety Solutions
SSIAFZIPMD |
Zip Lock Flu PPE Kit | 7 to 8, Medium |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
19-160-971
![]() |
Safety Solutions
SSIAFZIPLG |
Zip Lock Flu PPE Kit | 8 to 9, Large |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
19-160-969
![]() |
Safety Solutions
SSIAFDUFXXL |
Duffel Flu Ppe Kit | 9 to 10, 2XL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
19-160-968
![]() |
Safety Solutions
SSIAFDUFXL |
Duffel Flu Ppe Kit | 9 to 10, XL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
19-160-973
![]() |
Safety Solutions
SSIAFZIPXXL |
Zip Lock Flu PPE Kit | 9 to 10, 2XL |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. Part Not Found |
N/A |
Description
Contains personal protection and safety items for both those assisting persons exposed to influenzae and those involved in avian eradication activities. Respiratory protection meets CDC/NIOSH guidelines. Available in four sizes based on size-specific PPE products.
Specifications
- Contents are sealed inside a water-resistant zip lock bag
- Kits are also available with a sturdy, 12 x12 x 24 in. duffel bag for easy storage and transport of bagged components
- Breathable coverall with hood, elastic wrists, elastic ankles, and integrated booties for fast donning and excellent aerosol- and liquid-proof barrier protection
- Donning/Doffing instruction card provides Flu-specific information
Order Info
Duffel measures 24L x 12W x 12 in.H
1These kits have been assembled to help users reduce the risk of contracting infectious disease. Fit-testing, training, and use as recommended by manufacturers will increase the effectiveness of this equipment. However, there can be no assurance that use of the equipment will prevent the contraction of infectious disease. Protective apparel is consistent with CDC guidance for personal protection equipment. (See “Interim Recommendations for Infection Control in Health-Care Facilities Caring for Patients with Known or Suspected Avian Influenza,” U. S. Centers for Disease Control and Prevention, 5/21/04; and “Interim Guidance for Protection of Persons Involved in U. S. Avian Influenza Outbreak Disease Control and Eradication Activities,” U. S. Centers for Disease Control and Prevention, 2/17/04. www.cdc.gov.)
Specifications
CDC/NIOSH | |
Helps protect first responders, first receivers, and the private sector from exposure to influenza1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.