Learn More
Safety Solutions Flu PPE Kits
Helps protect first responders, first receivers, and the private sector from exposure to influenza1
Manufacturer:  Safety Solutions, Inc.Supplier Diversity Partner SSIAFZIPMD
 View more versions of this product
Catalog No. 19-160-970
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CDC/NIOSH
1 Pair safety goggles, side vent eye and face protection, 3M N95 disposable mask, model 1870 respiratory protection, 1 pair 4mil disposable nitrile gloves, powder-free, hand protection, 6.8mil nexgen coverall body protection, 1 pair latex boot covers foot protection, 2 dupont broad spectrum wipes, 4mil biohazard bag, donning/doffing instruction card safety supplies,
Zip Lock Flu PPE Kit
Helps protect first responders, first receivers, and the private sector from exposure to influenza1
7 to 8, Medium