Safety Solutions Flu PPE Kits
Helps protect first responders, first receivers, and the private sector from exposure to influenza1
Manufacturer: Safety Solutions, Inc.Supplier Diversity Partner SSIAFZIPXXL
 View more versions of this product
Catalog No. 19-160-973
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance CDC/NIOSH
    For Use With (Application) Helps protect first responders, first receivers, and the private sector from exposure to influenza1
    Includes 1 Pair safety goggles, side vent eye and face protection, 3M N95 disposable mask, model 1870 respiratory protection, 1 pair 4mil disposable nitrile gloves, powder-free, hand protection, 6.8mil nexgen coverall body protection, 1 pair latex boot covers foot protection, 2 dupont broad spectrum wipes, 4mil biohazard bag, donning/doffing instruction card safety supplies
    Size 9 to 10, 2XL
    Product Type Zip Lock Flu PPE Kit
  • Description & Specifications
Related Products
Product Content Feedback