Safety Solutions Occupational Flu PPE Kits
Helps protect you from exposure to influenza
Manufacturer: Safety Solutions, Inc.Supplier Diversity Partner SSIFLUINTXL
 View more versions of this product
Catalog No. 19-161-782
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) Assists person exposed to influenza
    Size X-Large
    Product Type Flu personal protection equipment kit
    Includes 1 Pair indirect vent goggles, 2 pair Nitrile gloves, 2 N95 respirators, 6 anti-bacterial handwipes, 1 biohazard bag, 1 don/doff instruction card
    Contains personal protection and safety items for those assisting persons exposed to influenza. Respiratory protection meets CDC/NIOSH guidelines. Available in three sizes (medium, large, and X-large) based on size-specific PPE products. All contents are sealed inside a water-resistant zip-lock bag.
    These kits have been assembled to help users reduce the risk of contracting influenza. Fit testing, training, and use as recommended by manufacturers will increase the effectiveness of this equipment. However, there can be no assurance that use of the equipment will prevent the contraction of infectious disease. Protective apparel is consistent with CDC guidance for personal protection equipment. (Refer to publications: “Interim Recommendations for Infection Control in Health-Care Facilities Caring for Patients with Known or Suspected Avian Influenza,” U. S. Centers for Disease Control and Prevention, 5/21/04; and “Interim Guidance for Protection of Persons Involved in U. S. Avian Influenza Outbreak Disease Control and Eradication Activities,” U. S. Centers for Disease Control and Prevention, 2/17/04. www.cdc.gov.)
  • Description & Specifications
Related Products
Product Content Feedback