Safety Solutions Smart-Strip Personal Protective Equipment Kits
Manufacturer: Safety Solutions, Inc.Supplier Diversity Partner PPEF01L
 View more versions of this product
Catalog No. 19-130-0310C
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Includes Tychem F coveralls - Size L, Nitrile inner and Butyl outer gloves, durable chemical overboots, chemical tape, custom ID tag, instruction card, decon kit, the Hazmat smart-strip, prepackaged in a durable backpack or duffel bag
    Product Type Personal protective equipment kit
    For Use With (Application) Protection for a broad range of chemicals
  • Description & Specifications
Related Products
Product Content Feedback