Learn More
PIP™ Safety Visor
PIP™ Safety Visor is ideal for general grinding applications.
Manufacturer:  PIP™ 251017214
Catalog No. 19-046-412
Add to cart
Description
Description
  • Made with virgin resin, which leads to better uniformity of the viewing area
  • Universal hole pattern will fit most headgear and hard hat adapters in the market
  • Ships with masking on both sides to prevent scratching
  • Meets ANSI Z87.1 Requirements
Specifications
Specifications
Clear
Clear
PETG
Universal fit
0.040 in.
Aluminum binding
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Warning - Prop 65 - Reproductive Harm