Learn More
MCR Safety SafetyToes™ Slipp-R™ Steel-Toe Safety Overshoes
100% rubber steel-toe safety overshoes feature an integrated steel toe, providing safety and durability.
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
19-825-560
|
MCR Safety
87008 |
Various | Includes the most popular size range | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
19-825-556
|
MCR Safety
87001 |
Pink | 2X-Small, 6-6.5M, W7-7.5 | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
19-825-559
|
MCR Safety
87007 |
Green | 2X-Large, M14, W15.5 | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
19-825-558
|
MCR Safety
87006 |
Blue | X-Large, M12-1/2, W14 | Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | ||||
Description
U.S. Safety SafetyToes Slipp-R Steel-Toe Safety Overshoes resist slips and cuts, keeping you on your toes and safer on the job.
- Flexible at all temperatures and easy to slip on
- Won't stiffen or crack
- Resistant to chemicals, acids and animal fats
- 100% waterproof
- Nonmarking
Compliance
Meet OSHA requirements for toe protection; 29 CFR 1910.136(b)(1). Passes ASTM F 2413-05 Paragraph 5.1.4.1(1) Impact Test and Paragraph 5.2.4.1(1) Compression Test in accordance with ASTM F 2412-05 Standard Test Method for Foot Protection.
Specifications
Rubber |
Product should not be used where metatarsal protection is required or where there is potential for electric shocks or objects piercing the sole.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.