Learn More
Selleck Chemical LLCSupplier Diversity Partner Safinamide Mesylate 10mg 202825-46-5 PNU-151774E,FCE28073  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S147210MG
Safinamide Mesylate (PNU-151774E, FCE28073) is mesylate salt of Safinamide, selectively and reversibly inhibits MAO-B with IC50 of 98 nM, exhibits 5918-fold selectivity against MAO-A. Phase 3.
Catalog No. 50-634-5