Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Safranine O (C.I. 50240), For Microscopy, Certistain™, MilliporeSigma™

Click to view available options
Quantity:
25g
477-73-6
C20H19ClN4
350.85
MFCD00011759
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
C.I. 50240
2723800
2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
477-73-6
C20H19ClN4
350.85
MFCD00011759
QRYAEWIQIBAZOJ-UHFFFAOYSA-N
C.I. 50240
2723800
2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
Appearance Red brown solid
Formula Weight 350.85
Physical Form Solid
Odor Odorless
Packaging Glass bottle
Quantity 25g
Chemical Name or Material Safranine O