Sage P2 Safety Plus High-Risk Latex Gloves
Manufacturer: Moore Medical Corp 53661
 View all options for this product
Catalog No. 19-815-390
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Hand Ambidextrous
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Material Latex
    Powdered No
    Size 9 to 9.5 (XL)
    Thickness 15mil (fingertip)
    Certifications NFPA 1999, OSHA and ASTM
  • Description & Specifications
Related Products
Product Content Feedback