Learn More
Sage Science™ BluePippin™ 2% Agarose Gel Cassettes
For size selections between 100bp and 600bp. Sage Science™ BluePippin™ 2% Agarose Gel Cassettes are commonly used for Illumina paired-end library construction, Ion Torrent template prep, and ChIP applications.
$671.00
Products 1
Catalog Number Mfr. No. Quantity Product Type Specs Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Specs Price Quantity & Availability  
12-100-519
Encompass
View Documents
Sage Science™
BEF2010
10 Pack Agarose Gel Cassette
Pack of 10 for $671.00
Add to cart
Description
Description
  • Includes sufficient reagents to run all cassettes in pack
  • Dye free cassettes, with internal or external marker, runs four or five samples per cassette
  • Band capture cassettes contain dye and external marker, runs four samples per cassette
Order Info
Cassettes for EpiCentre ScriptSeq complete kits available separately (Cat. Nos. 12-100-543 and 12-100-521).