Learn More
SAGE1 Mouse anti-Human, Clone: 5C1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524474
Catalog No. PIMA524474
Add to cart
Description
Description
Peptide Sequence: PLPSNALSTV LPGLAYLATA DMPAMSTRDQ HATIIHNLRE EKKDNSQPTP DNVLSAVTPE LINLAGAGIP PMSTRDQYAT VNHHVHEARM ENGQRKQDN
This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas.
Specifications
Specifications
SAGE1
Monoclonal
Unconjugated
SAGE1
Liquid
SAGE1
SAGE1 (NP_061136, 357 a.a. ∽ 455 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
5C1
PBS with no preservative; pH 7.4
Q9NXZ1
SAGE1; SAGE
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
55511