Learn More
Saint-Gobain ALUNDUM™ Extraction Thimbles
Wash, ignite, and use repeatedly
Manufacturer:  Saint-Gobain AN485 5811
 View more versions of this product
Catalog No. 09-661C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Used for all kinds of extractions except strong alkalis
  • ALUNDUM AN889 thimbles (Series 09-660) have medium porosity, retain particles approximately 5µm and larger
  • ALUNDUM AN485 thimbles (Series 09-661) have coarse porosity for faster extraction, less retentivity; retain particles approximately 20µm and larger
  • 34 and 45mm diameter thimbles in each series have round bottoms; all other sizes have flat bottoms
†With round bottoms.
Specifications
Specifications
Flat
3cm
3.14 in.
20μm
Extraction Thimble
1.18 in.
Used for all kinds of extractions except strong alkalis
8cm
Coarse
Alundum™